Koehler Constant-Temperature Viscosity Baths
Save time – seven viscometer holders and seven bath timers allow multiple measurements at the same time
  • Motorized stirrer provides uniform temperature control and thorough circulation without turbulence
Baths feature PID microprocessor controller to deliver precise temperature control, with ASTM specified tolerances throughout the operating temperature range. Dual digital displays show set point time and actual temperature. Timers display in 0.01-second resolution and have ±0.01% accuracy.
Controller automatically displays test results in efflux time and viscosity units when timers stop.
Controller automatically shuts off power when there is an overtemperature condition or if the primary probe is disconnected. The integrated low-level sensor interrupts operations if the bath liquid does not fill to the proper level, and shuts off power if the liquid falls below the level.
Bath chamber is a clear Pyrex® tank enclosed in a polyester-epoxy finished steel housing. Top plate holds the viscometers and is made of stainless steel for easy cleaning. Front viewing window reduces distortion. Glare-free fluorescent lights in the bath and a background baffle enhance the view of the viscometers. Baths rest on adjustable leveling feet.
All models conform to ASTM D445, D2170, D6074, D6158; IP 71, 319; ISO 3104; DIN 51550; FTM 791-305.
Call For Price
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Interface Power (VAC) Reservoir Capacity (L)
Availability Pricing
Koehler K23790 Constant-Temperature Viscosity Bath, 22 L, 230 VAC WW-98944-05 Mfr # K23790
COMPARE
None 230 22