Cole-Parmer® TELOS® Supported Liquid Extraction (SLE) Columns
Perfectly suited for automation procedures
  • Efficient extraction
  • High analyte recovery with no emulsion formation
Use supported liquid extraction columns when you need to extract samples quickly and effectively. Columns are packed with a diatomaceous earth, a neutral matrix, to provide support to absorb the aqueous sample. Once the aqueous sample is fully absorbed, a water immiscible organic solvent is used to elute the analytes. High analyte recoveries are obtained without any offline steps such as protein precipitation, and sample extracts are free of proteins and phospholipids. A hydrophilic top frit allows the sample to pass through while minimizing any blockage.
12,400.00 - ₹22,300.00INR / Pkg of 8
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Applications Bed Mass (g)
Availability Pricing
COMPARE
150 Neutral matrix 10
₹13,100.00INR / Pkg of 8
COMPARE
25 Neutral matrix 2.5
₹22,300.00INR / Pkg of 20
COMPARE
150 Neutral matrix (NM) 545 10
₹12,400.00INR / Pkg of 8
COMPARE
25 Neutral matrix (NM) 545 2.5
₹22,300.00INR / Pkg of 20