Cole-Parmer® TELOS® SPE-Format Silica Flash Chromatography Columns
Ideal for first stage clean-up columns post synthesis using a vacuum manifold
  • Excellent for purification of a wide range of synthetic and natural compounds
Easily connect to flash chromatography system using the Luer-slip connection. Use columns on a rack or stand using gravity flow or with vacuum processing stations. All columns are made from polypropylene and come with polyethylene frits.
6,400.00 - ₹18,100.00INR / Pkg of 20
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (g)
Availability Pricing
COMPARE
2
₹6,400.00INR / Pkg of 20
COMPARE
25 5
₹13,100.00INR / Pkg of 20
COMPARE
70 10
₹13,000.00INR / Pkg of 16
COMPARE
70 20
₹15,300.00INR / Pkg of 16
COMPARE
70 50
₹14,600.00INR / Pkg of 8
COMPARE
150 70
₹18,100.00INR / Pkg of 8