Cole-Parmer® TELOS® SPE-Format C18 Flash Chromatography Columns
Ideal for first stage clean-up columns post synthesis using a vacuum manifold
  • Bonded octadecyl stationary phase offers reverse phase flash capabilities
  • Has a pore size of 60Å and a carbon loading of ≈18%
Easily connect to flash chromatography system using the Luer-slip connection. Use columns on a rack or stand using gravity flow or with vacuum processing stations. All columns are made from polypropylene and come with polyethylene frits.
29,800.00 - ₹75,600.00INR / Pkg of 20
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (g)
Availability Pricing
COMPARE
25 5
₹29,800.00INR / Pkg of 20
COMPARE
70 20
₹75,600.00INR / Pkg of 16
COMPARE
150 50
₹58,300.00INR / Pkg of 8