Cole-Parmer® TELOS® SPE Blank Fixed 96-Well Microplates
Pack wells with custom sorbents
  • Optimized for the sample processing and extraction of small volume fluids
  • Use with vacuum or positive pressure
Individual well is designed to allow maximum interaction between the analyte and sorbent and ensure elution volumes are kept to a minimum. Fill with your own custom sorbent to meet your application. Each well is fitted with a polyethylene frit.
7,900.00 - ₹8,100.00INR / Pkg of 1
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fritted Disk
Availability Pricing
COMPARE
20 µm
₹7,900.00INR / Pkg of 1
COMPARE
10 µm
₹8,100.00INR / Pkg of 1