Cole-Parmer® TELOS® Silica Polar SPE Chromatography Columns
Extract polar analytes from nonpolar organic solvents
  • Compatible with manual and automated sample protocols
Use TELOS polar (normal phase) silica SPE columns to retain the analytes of interest or to remove polar interferences from the sample. Silica sorbent has a pore diameter of 70Å. All columns are made of polypropylene and feature Luer-slip outlet.
8,900.00 - ₹20,500.00INR / Pkg of 30
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
6 1000
₹10,100.00INR / Pkg of 30
COMPARE
15 2000
₹10,500.00INR / Pkg of 20
COMPARE
25 5000
₹13,100.00INR / Pkg of 20
COMPARE
1 100
₹20,500.00INR / Pkg of 100
COMPARE
3 100
₹9,300.00INR / Pkg of 50
COMPARE
3 200
Call For Price
COMPARE
3 500
₹10,300.00INR / Pkg of 50
COMPARE
6 500
₹8,900.00INR / Pkg of 30
Showing 1- 8 of 8