Cole-Parmer® TELOS® Silica Flash Chromatography Columns with Luer-lok® inlet and outlet
Use with SP and Idolera™ systems from Biotage®
  • Optimized column geometry provides maximum recovery
  • Excellent for purification of a wide range of synthetic and natural compounds
Make connections to flash chromatography instruments quick, easy, and secure with Luer-lok connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.
15,300.00 - ₹21,100.00INR / Pkg of 20
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Bed Mass (g)
Availability Pricing
COMPARE
12
₹21,100.00INR / Pkg of 20
COMPARE
20
₹21,100.00INR / Pkg of 20
COMPARE
40
₹16,000.00INR / Pkg of 10
COMPARE
80
₹15,300.00INR / Pkg of 5
COMPARE
120
₹16,600.00INR / Pkg of 5