This product is now discontinued, View Related Products
Cole-Parmer® TELOS® Silica-Based SPE Populated-Well Microplates
Comes with 96 wells already in place yet allow the flexibility to remove any unwanted wells
  • Optimized for sample processing and extraction of small volume biological fluids
  • Use with vacuum or positive pressure
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® Silica-Based SPE populated-well microplates allow for maximum recovery with minimal sample dilution and reduced evaporation time. Select endcapped sorbents for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. Use C18 (AQ) with samples with large volumes of water.
30,900.00 - ₹33,800.00INR / Pkg of 1
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Sorbent Bed Mass (mg) End Cap
Availability Pricing
COMPARE
C18 5 No
Call For Price
COMPARE
C18 10 No
₹33,800.00INR / Pkg of 1
COMPARE
C18 10 Yes
₹33,800.00INR / Pkg of 1
COMPARE
C18 (AQ) 5 Yes
₹33,200.00INR / Pkg of 1
COMPARE
C18 (AQ) 10 Yes
₹30,900.00INR / Pkg of 1