Cole-Parmer® TELOS® Silica-Based SPE Fixed 96-Well Microplates
Optimized for sample processing and extraction of small volume biological fluids
  • Select from three main retention mechanisms: nonpolar, ion exchange, or polar
  • Use with vacuum or positive pressure
The internal diameter, frit design, and sorbent material of the Cole-Parmer® TELOS® Silica-Based SPE fixed 96-well microplates allow for maximum recovery with minimal sample dilution and reduced evaporation time. Select endcapped sorbents for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. C4 (WP) sorbents with average pore size of 300 Å are ideal for compounds with molecular weight >10,000.
21,300.00 - ₹35,900.00INR / Pkg of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sorbent Bed Mass (mg) End Cap
Availability Pricing
COMPARE
Silica 100 No
₹26,600.00INR / Pkg of 1
COMPARE
Silica 350 No
₹29,500.00INR / Pkg of 1
COMPARE
NH2 50 No
₹21,300.00INR / Pkg of 1
COMPARE
C18 50 No
₹33,800.00INR / Pkg of 1
COMPARE
C18 100 No
₹35,900.00INR / Pkg of 1
COMPARE
C18 50 Yes
₹31,400.00INR / Pkg of 1
COMPARE
SAX 50 No
₹27,400.00INR / Pkg of 1
Showing 1- 7 of 7