Cole-Parmer® TELOS® SCX Ion Exchange SPE Columns
Extract basic analytes with a positive charge from aqueous samples
  • Features Benzenesulfonic acid sorbent
TELOS SCX (benzenesulfonic acid) columns have average pore diameter of 70 Å with 0.6 meq/g exchange capacity. All columns are made of polypropylene and come with Luer-slip outlet. Ionizable basic analytes with pK range 8 to 12.
21,700.00 - ₹32,800.00INR / Pkg of 30
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
6 1000
₹32,800.00INR / Pkg of 30
COMPARE
3 200
₹21,700.00INR / Pkg of 50
COMPARE
3 500
₹32,000.00INR / Pkg of 50