Cole-Parmer® TELOS® SCX-2 Ion Exchange SPE Columns
Extract basic analytes with a positive charge from aqueous samples
  • Features Propylsulfonic acid sorbent
TELOS SCX-2 (propylsulfonic acid) allows elution with an aqueous solvent, providing a cleaner final extract than SCX—ideal for isolating and purifying basic compounds following organic synthesis. SCX-2 columns have average pore diameter of 70 Å with 0.3 meq/g exchange capacity. All columns are made of polypropylene and come with Luer-slip outlet. Ionizable basic analytes with pK range 8 to 12.
38,000.00 - ₹258,000.00INR / Pkg of 30
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (g)
Availability Pricing
COMPARE
6 1
₹38,000.00INR / Pkg of 30
COMPARE
25 5
₹59,100.00INR / Pkg of 20
COMPARE
70 10
₹92,900.00INR / Pkg of 16
COMPARE
150 25
₹106,000.00INR / Pkg of 8
COMPARE
150 50
₹258,000.00INR / Pkg of 8