Cole-Parmer® TELOS® PS-DVB and PS-DVB ENV SPE Columns
Ideal for polar water-soluble analyte extraction from aqueous samples
  • Manufactured with PS-DVB (polystyrene divinylbenzene resin) and PS-DVB ENV (hydroxylated polystyrene divinylbenzene resin) solvents
PS-DVB (polystyrene divinylbenzene resin) and PS-DVB ENV (hydroxylated polystyrene divinylbenzene resin) are ideal for polar water-soluble analyte extraction from aqueous samples. PS-DVB ENV has a very high surface area, offering excellent retention of very polar analytes including phenols and phenoxyacid herbicides from environmental samples and drug metabolites from biological fluids. Columns are made of polypropylene and feature Luer-slip outlet.
20,700.00 - ₹46,700.00INR / Pkg of 30
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sorbent Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
PS-DVB (ENV) 6 200
₹22,100.00INR / Pkg of 30
COMPARE
PS-DVB 6 1000
₹46,700.00INR / Pkg of 30
COMPARE
PS-DVB 3 100
₹20,700.00INR / Pkg of 50
COMPARE
PS-DVB 6 500
₹26,200.00INR / Pkg of 30