Cole-Parmer® TELOS® Phase Separator Columns
Quickly and easily separate aqueous and chlorinated solvents
  • Gravity flow filtration process, no vacuum required
  • No need for separating funnels and rewashing glassware
Simply apply aqueous and chlorinated solvents to the column for a quick separation. A special hydrophobic frit allows the chlorinated solvent to pass through under gravity while preventing the aqueous layer from flowing though. The aqueous layer remains in the separator column, separating the two phases and allowing additional work up.
11,700.00 - ₹14,700.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL)
Availability Pricing
COMPARE
6
₹14,000.00INR / Pkg of 100
COMPARE
15
₹14,000.00INR / Pkg of 100
COMPARE
70
₹11,700.00INR / Pkg of 50
COMPARE
150
₹14,700.00INR / Pkg of 25