Cole-Parmer® TELOS® NH2 Flash Chromatography Columns with Luer-lok® inlet and Luer-slip outlet
Use with systems from ISCO® and Buchi®
  • Optimized column geometry provides maximum recovery
  • Use to separate primary, secondary, and tertiary amines
  • NH2 shows better peak shapes with polar and basic compound—ideal for carbohydrates
Make connections to flash chromatography instruments quick and easy with Luer connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.
25,700.00 - ₹134,000.00INR / Pkg of 20
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Bed Mass (g)
Availability Pricing
COMPARE
4
₹25,700.00INR / Pkg of 20
COMPARE
12
₹42,700.00INR / Pkg of 20
COMPARE
20
₹90,900.00INR / Pkg of 20
COMPARE
120
₹134,000.00INR / Pkg of 5