Cole-Parmer® TELOS® neo™ PRP SPE Columns
Nonpolar SPE columns for acidic, neutral, and basic analyte extraction
  • High surface area and capacity ensure highly reliable retention characteristics
  • No secondary interactions from silanol groups for accurate results
  • Higher sample loading due to high surface area
Recover clean extracts and shorten the time spent on method development using TELOS neo PCR (polar-modified reversed phase) SPE columns. Reversed phase (nonpolar) SPE remains a popular due to its applicability to a wide range of compounds.
High surface area and capacity allow the use of smaller sorbent masses, leading to reduced elution volumes and evaporation/reconstitution steps. Polymeric sorbents are water-wettable and not affected by drying out, providing better flow characteristics, retention, and elution.
16,900.00 - ₹38,800.00INR / Pkg of 50
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
3 100
₹30,400.00INR / Pkg of 50
COMPARE
1 30
₹23,400.00INR / Pkg of 100
COMPARE
6 500
₹38,800.00INR / Pkg of 30
COMPARE
3 60
₹19,500.00INR / Pkg of 50
COMPARE
6 200
₹16,900.00INR / Pkg of 30