Cole-Parmer® TELOS® neo™ PAX SPE Columns
Mixed-mode strong anion exchange columns for acidic analyte extraction
  • High surface area and capacity ensure highly reliable retention characteristics
  • No secondary interactions from silanol groups for accurate results
  • Higher sample loading due to high surface area
Recover clean extracts and shorten the time spent on method development using TELOS neo polymeric SPE columns. High surface area and capacity allow the use of smaller sorbent masses, leading to reduced elution volumes and evaporation/reconstitution steps. Polymeric sorbents are water-wettable and not affected by drying out, providing better flow characteristics, retention, and elution. Columns provide high and reproducible recoveries for a wide range of analytes, including acidic, neutral, basic, and multi-charged analytes.
14,300.00 - ₹22,800.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
1 30
₹22,800.00INR / Pkg of 100
COMPARE
6 200
₹20,000.00INR / Pkg of 30
COMPARE
3 30
₹14,300.00INR / Pkg of 50
COMPARE
3 60
₹19,300.00INR / Pkg of 50