Cole-Parmer® TELOS® Multilayer SPE Columns
Achieve superior clean up when conducting multiresidue analysis
  • Save money by extracting multiple suites with a single column
Multilayered SPE columns are designed for efficient extraction of multi-analyte suites. Layers of different SPE sorbents in a single SPE column allow a broader range of analytes to be extracted than is possible with a single sorbent. Each column features 20 µm frits at the top, bottom, and between sorbent layers. Columns feature Luer-slip outlet.
19,000.00 - ₹25,500.00INR / Pkg of 30
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sorbent Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
SAX/PSA 6 1000
₹19,000.00INR / Pkg of 30
COMPARE
Na₂SO₄/MgO₃Si 6 2000
₹25,500.00INR / Pkg of 30
COMPARE
Amino/octadecyl (PAH) 6 1500
₹19,000.00INR / Pkg of 30
COMPARE
PSA/C18 6 500
Call For Price
COMPARE
C18/ENV (endcapped) 6 700
₹22,300.00INR / Pkg of 30