This product is now discontinued, View Related Products
Kinesis® TELOS® HPLC and UHPLC Column Accessories
Help extend the life of your columns
  • Optimize performance of your Kinesis® TELOS® HPLC and UHPLC columns
Support your columns with optional guard cartridges (and holder), available packed with identical packing material.
23,600.00 - ₹76,800.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Compatible With Description
Availability Pricing
COMPARE
10 mm ID C18 columns TELOS® Guard Cartridge, C18, Preparative 5 µm, 10 mm ID; 1/EA
₹66,800.00INR / Each
COMPARE
Guard cartridges TELOS® Guard Cartridge Holder; 1/EA
₹23,600.00INR / Each
COMPARE
4.6 mm ID silica HILIC columns TELOS® Guard Cartridge, Silica HILIC, 5 µm, 4.6 mm ID; 5/PK
₹62,900.00INR / Pkg of 5
COMPARE
4.6 mm ID polar C18 columns TELOS® Guard Cartridge, C18, Polar, 5 µm, 4.6 mm ID; 5/PK
₹62,900.00INR / Pkg of 5
COMPARE
4.6 mm ID C8 columns TELOS® Guard Cartridge, C8, 5 µm, 4.6 mm ID; 5/PK
₹69,600.00INR / Pkg of 5
COMPARE
4.6 mm ID cyano columns TELOS® Guard Cartridge, Cyano, 5 µm, 4.6 mm ID; 5/PK
₹62,900.00INR / Pkg of 5
COMPARE
4.6 mm ID C18 columns TELOS® Guard Cartridge, C18, 5 µm, 4.6 mm ID; 5/PK
₹76,800.00INR / Pkg of 5
Showing 1- 7 of 7