This product is now discontinued, View Related Products
Cole-Parmer® TELOS® H-CX (Hydrophobic Cation) SPE Columns
Optimized for basic drug extraction from biological fluids
  • Manufactured with H-CX (hydrophobic/cation) sorbent
H-CX (hydrophobic/cation) is a mixed-mode sorbent optimized for basic drug extraction from biological fluids. The dual retention mechanism allows an extensive interference elution regime, removing salts, polar interferences, peptides, and proteins prior to analyte elution. H-CX is a commonly use sorbent for many drugs of abuse applications. Columns are made of polypropylene and feature Luer-slip outlet.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
1 130
Call For Price
COMPARE
3 130
Call For Price
COMPARE
3 300
Call For Price
COMPARE
3 500