Cole-Parmer® TELOS® Graphitized Carbon SPE Columns
Ideal for polar water-soluble analyte extraction from aqueous sample
  • Manufactured with graphitized carbon sorbent
Graphitized carbon sorbent is a great for polar water-soluble analyte extraction from aqueous samples. It has an extremely high affinity for a very wide range of analytes using a nonpolar retention mechanism. Columns are made of polypropylene and feature Luer-slip outlet.
21,100.00 - ₹88,600.00INR / Pkg of 30
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
6 1000
₹88,600.00INR / Pkg of 30
COMPARE
1 50
₹31,900.00INR / Pkg of 100
COMPARE
6 200
₹21,100.00INR / Pkg of 30