Cole-Parmer® TELOS® Flash Chromatography Bulk Sorbent Media
High-purity adsorbents for nonpolar, ion exchange, and polar retention mechanisms
  • Use when samples need to be pre-absorbed onto loose sorbent material
  • Suitable for dry loading and packing home-built columns
The TELOS bulk sorbents for flash chromatography dry loading are available in silica, NH2 (bonded aminopropyl), C18 (octadecyl), and NM (diatomaceous earth). Silica, NH2, and C18 have 40 to 60 µm particle size and pore size of 60 Å. These sorbent materials allow samples to be pre-absorbed before loading onto the top of a flash column:
1. Pre-absorb sample in one of the TELOS bulk sorbent material.
2. Solvent is removed.
3. The resulting sample-sorbent blend is applied to the sample loading device or right on the top frit of the flash column.
25,200.00 - ₹338,000.00INR / Pkg of 1
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Sorbent Media Mass (g)
Availability Pricing
COMPARE
Silica 1000
₹25,200.00INR / Pkg of 1
COMPARE
Silica 5000
Call For Price
COMPARE
NH₂ 100
₹25,400.00INR / Pkg of 1
COMPARE
NM (diatomaceous earth) 100
₹79,000.00INR / Pkg of 1
COMPARE
NM (diatomaceous earth) 1000
₹338,000.00INR / Pkg of 1
COMPARE
C18 1000
₹30,000.00INR / Pkg of 1
COMPARE
C18 5000
₹314,000.00INR / Pkg of 1
Showing 1- 7 of 7