Cole-Parmer® TELOS® Fittings
Optimize performance of your TELOS chromatography columns
  • Choose from a wide range of accessories for flash chromatography dry loading, filtration, and large volume processing
Provide a secure seal for both the inlet and outlet of TELOS flash columns with the luer cap and plug. TELOS column luer cap can be used in conjunction with the appropriate size column for either room temperature reactions or short-term compound storage.
Easily and quickly adapt 1-, 3-, or 6-mL TELOS SPE columns the column adapter.
3,200.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Color Compatible With Description
Availability Pricing
COMPARE
Black Flash chromatography columns TELOS Luer Cap
₹3,200.00INR / Pkg of 100
COMPARE
Clear 1 mL, 3 mL, or 6 mL columns TELOS Column Adapters for 1-, 3- and 6 mL Columns
₹3,200.00INR / Pkg of 10