Cole-Parmer® TELOS® Filtration Columns
Comes with frits for simple filtration and sample processing
  • Select columns with polyethylene (PE) or PTFE frits
  • PE frit models available with single- or double-frit configuration
Total volumes for the columns range from 1 to 150 mL. Columns come with either polyethylene or PTFE frits in 5, 10, or 20 µm pore sizes. PE frits area available in single- or double-fritted configurations. Double frit columns have two frits one directly on top of the other to provide better filtration and cleaner sample. Use caps for room-temperature reactions and to easily transport columns.
12,300.00 - ₹32,800.00INR / Pkg of 100
17 variations of this product are available.
See All Product Options Below
17 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Column Particle Size (µm) Fritted Disk
Availability Pricing
COMPARE
15 20 Double, Polyethylene
₹16,400.00INR / Pkg of 100
COMPARE
70 10 Single, Polyethylene
₹13,100.00INR / Pkg of 50
COMPARE
15 10 Single, PTFE
₹24,600.00INR / Pkg of 100
COMPARE
6 20 Double, Polyethylene
₹13,400.00INR / Pkg of 100
COMPARE
25 20 Double, Polyethylene
₹21,300.00INR / Pkg of 100
COMPARE
70 20 Double, Polyethylene
₹19,700.00INR / Pkg of 50
COMPARE
6 5 Single, PTFE
₹18,500.00INR / Pkg of 100
COMPARE
15 5 Single, PTFE
₹24,600.00INR / Pkg of 100
COMPARE
150 20 Double, Polyethylene
₹13,900.00INR / Pkg of 25
COMPARE
70 10 Double, Polyethylene
₹14,800.00INR / Pkg of 50
COMPARE
25 10 Single, PTFE
₹32,800.00INR / Pkg of 100
COMPARE
3 20 Double, Polyethylene
₹15,600.00INR / Pkg of 100
COMPARE
15 10 Double, Polyethylene
₹14,500.00INR / Pkg of 100
COMPARE
15 10 Single, Polyethylene
₹13,100.00INR / Pkg of 100
COMPARE
1 20 Double, Polyethylene
₹12,300.00INR / Pkg of 100
COMPARE
10 20 Double, Polyethylene
₹20,700.00INR / Pkg of 100
COMPARE
6 10 Single, PTFE
₹18,600.00INR / Pkg of 100
Showing 1- 17 of 17