Cole-Parmer® TELOS® Dry Loading Columns with Luer-lok® inlet and outlet
Use with SP and Idolera™ systems from Biotage®
  • Columns are prefitted with a bottom frit
Use dry loading columns with bulk sorbents to quickly and easily apply to flash chromatography system. Attach dry loading columns directly to the main flash columns to suit majority of reaction scales. Columns feature Luer-lok inlet and outlet and come prefitted with a bottom frit. Simply add the reaction mixture, place a top frit (included) in place, then screw the top in place.
11,600.00 - ₹15,600.00INR / Pkg of 20
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Bed Mass (g) Fritted Disk
Availability Pricing
COMPARE
20 20 µm
₹15,600.00INR / Pkg of 20
COMPARE
40 10 µm
₹11,600.00INR / Pkg of 10