Cole-Parmer® TELOS® C8 Nonpolar SPE Columns
Extract hydrophobic or polar organic analytes from aqueous matrices
  • Features C8 sorbent
TELOS nonpolar (reversed phase) SPE columns are available in a wide range of sorbents for extraction of hydrophobic and polar analytes. Select columns with endcaps for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. All columns are made of polypropylene and feature Luer-slip outlet. TELOS C8 has a pore size of 70 Å and average particle size of 50 µm.
18,400.00 - ₹23,900.00INR / Pkg of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg) End Cap
Availability Pricing
COMPARE
1 100 Yes
₹23,900.00INR / Pkg of 100
COMPARE
3 500 Yes
₹18,400.00INR / Pkg of 50