This product is now discontinued, View Related Products
Cole-Parmer® TELOS® C4 Nonpolar SPE Columns
Extract hydrophobic or polar organic analytes from aqueous matrices
  • Features C4 sorbent
TELOS nonpolar (reversed phase) SPE columns are available in a wide range of sorbents for extraction of hydrophobic and polar analytes. All columns are made of polypropylene and feature Luer-slip outlet. TELOS C4 has a pore size of 300 Å and average particle size of 50 µm.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
1 50
Call For Price
COMPARE
3 100