This product is now discontinued, View Related Products
Cole-Parmer® TELOS® C2 Nonpolar SPE Columns
Extract hydrophobic or polar organic analytes from aqueous matrices
  • Features C2 sorbent
TELOS nonpolar (reversed phase) SPE columns are available in a wide range of sorbents for extraction of hydrophobic and polar analytes. All columns are endcapped making them ideal for applications requiring minimal secondary silanol interactions to enhance compound retention and elution. Columns are made of polypropylene and feature Luer-slip outlet. TELOS C2 has a pore size of 70 Å and average particle size of 50 µm.
Call For Price
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Total Volume Capacity (mL) Bed Mass (mg) End Cap
Availability Pricing
COMPARE
1 100 Yes
Call For Price
COMPARE
3 100 Yes
Call For Price
COMPARE
3 200 Yes
Call For Price
COMPARE
3 500 Yes