Cole-Parmer® TELOS® C18 Flash Chromatography Columns with Luer-lok® inlet and outlet
Use with SP and Idolera™ systems from Biotage®
  • Optimized column geometry provides maximum recovery
  • Bonded octadecyl stationary phase offers reverse phase flash capabilities
  • Has a pore size of 60Å and a carbon loading of ≈18%
Make connections to flash chromatography instruments quick, easy, and secure with Luer-lok connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.
121,000.00INR / Pkg of 20
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Bed Mass (g)
Availability Pricing
COMPARE
23
₹121,000.00INR / Pkg of 20