Cole-Parmer® TELOS® C18 Flash Chromatography Columns with Luer-lok® inlet and Luer-slip outlet
Use with systems from ISCO® and Buchi®
  • Optimized column geometry provides maximum recovery
  • Bonded octadecyl stationary phase offers reverse phase flash capabilities
  • Has a pore size of 60Å and a carbon loading of ≈18%
Make connections to flash chromatography instruments quick and easy with Luer connectors. Columns are made of polypropylene and have pressure ratings to support common flash chromatography back pressures.
27,300.00 - ₹196,000.00INR / Pkg of 20
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Bed Mass (g)
Availability Pricing
COMPARE
4
₹27,300.00INR / Pkg of 20
COMPARE
12
₹50,800.00INR / Pkg of 20
COMPARE
23
₹121,000.00INR / Pkg of 20
COMPARE
45
₹141,000.00INR / Pkg of 10
COMPARE
90
₹133,000.00INR / Pkg of 5
COMPARE
120
₹196,000.00INR / Pkg of 5
COMPARE
375
₹84,700.00INR / Pkg of 1
Showing 1- 7 of 7