Cole-Parmer® TELOS® Bulk SPE Media
Save money by purchasing high quality sorbents in bulk quantities
  • Choose from a wide range of SPE media for packing your own columns
Pack your column with a single sorbent or create your own multilayer column using bulk media. Sorbents come in 100 g or 1 kg bottles.
21,100.00 - ₹314,000.00INR / Pkg of 1
9 variations of this product are available.
See All Product Options Below
9 PRODUCT OPTIONS
Compareitem specifications
Sorbent End Cap Media Mass (g)
Availability Pricing
COMPARE
SCX-2 (C3H8O3S) No 100
₹70,400.00INR / Pkg of 1
COMPARE
Graphitized carbon No 100
₹169,000.00INR / Pkg of 1
COMPARE
PSA (primary-secondary amine) No 100
₹30,200.00INR / Pkg of 1
COMPARE
Alumina—Neutral (AL-N) No 100
₹21,100.00INR / Pkg of 1
COMPARE
C18 (octadecyl) No 100
₹40,100.00INR / Pkg of 1
COMPARE
C18 (octadecyl) Yes 100
₹39,100.00INR / Pkg of 1
COMPARE
C18 (flash octadecyl) No 1000
₹314,000.00INR / Pkg of 1
COMPARE
SCX (C6H6O3S) No 100
₹59,100.00INR / Pkg of 1
COMPARE
SAX (NR4+) No 100
₹59,100.00INR / Pkg of 1
Showing 1- 9 of 9