Kinesis® Short Thread TPX PMP/TPX PP Vials, 9 mm
Compatible with all autosamplers
  • TPX PMP has excellent chemical resistance and durability
Short thread vials have a wide opening (6.0 mm ID) with a unique thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
TPX PMP vials combine the robustness of TPX PMP with the benefits of glass or PP micro inserts. Insert sits slightly higher than the edge of the vial rim to ensure that the septum seals tightly. Order salinized vials to minimize adsorption of polar compounds, including amino acids, proteins, and phenols to the inner surface of the vial.
53,700.00 - ₹159,000.00INR / Pkg of 1000
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features Tube Color Volume (mL)
Availability Pricing
COMPARE
Fused PP micro insert Clear 0.3
₹53,700.00INR / Pkg of 1000
COMPARE
Fused glass micro insert Clear 0.2
₹84,900.00INR / Pkg of 1000
COMPARE
Fused glass micro insert Amber 0.2
₹89,600.00INR / Pkg of 1000
COMPARE
Silanized, fused glass micro insert Amber 0.2
₹154,000.00INR / Pkg of 1000
COMPARE
Silanized, fused glass micro insert Clear 0.2
₹159,000.00INR / Pkg of 1000