Kinesis® Screw Cap Vial Kits
Vial kits combine vials and caps with septa for your convenience
  • Resealable box helps prevent contamination during use
  • Constructed with silicone/PTFE septa for contaminant free sampling
  • 2-mL size ideal for chromatography applications
Vials are available in clear or amber glass with label and filling lines. Amber vials are ideal for light sensitive samples.
UltraClean silicone/PTFE septa are optimized for gas chromatography and headspace analysis. The pre-slit septa prevent a vacuum forming inside the vial.
11,400.00INR / Pkg of 1000
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description Cap Type Tube Color
Availability Pricing
COMPARE
Crimp Neck Vial and Cap Kit, HPLC/GC Certified Amber Glass Vials, 11 mm, UltraClean ™ Silicone/PTFE Septa; 1000/pk Crimp neck Amber
₹11,400.00INR / Pkg of 1000