This product is now discontinued, View Related Products
Cole-Parmer® Scintillation Glass Vials and Caps
Ensure accuracy with low-potassium glass vials in your liquid scintillation systems
  • Type I low-potassium neutral glass withstands most chemicals
  • Choose amber glass for light sensitive samples
Low-potassium borosilicate glass scintillation vials provide low activity counts and excellent light transmission. By minimizing the potassium levels in our glass formulations, we can ensure reliable, consistent results in scintillation counting. These flat-bottomed vials come with white polypropylene foil-lined urea screw caps.
59,700.00 - ₹68,900.00INR / Pkg of 500
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Tube Color Tube Material Volume (mL)
Availability Pricing
COMPARE
Clear Glass 20
₹68,900.00INR / Pkg of 500
COMPARE
Clear Glass 20
₹59,700.00INR / Pkg of 500