Cole-Parmer® TELOS® Polar SPE Cartridges
Extract polar analytes from nonaqueous matrices
  • Constructed of durable polypropylene
Use the TELOS polar (normal phase) SPE cartridges to retain analytes of interest or to remove polar interferences from the sample. Columns with Florisil® PR grade (magnesium silicate) sorbent have larger particle size than NH2 and are commonly used for lipid separation and pesticide extraction. All columns are made of polypropylene and feature female Luer-slip inlet and male Luer-slip outlet.
12,700.00 - ₹158,000.00INR / Pkg of 50
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Sorbent Total Volume Capacity (mL) Bed Mass (mg)
Availability Pricing
COMPARE
NH2 1 mL 600
₹12,700.00INR / Pkg of 50
COMPARE
₹158,000.00INR / Pkg of 50
COMPARE
Florisil® PR grade 1 mL 750
₹16,900.00INR / Pkg of 50
COMPARE
₹68,100.00INR / Pkg of 50