Kinesis KX Regenerated Cellulose Syringe Filters with Prefilter
Regenerated cellulose membranes are hydrophilic and ideal for particle removal from solvents and mobile phase filtration for HPLC
  • Improve removal of larger and unwanted buildup with a two-stage filter
  • Increase flow rate with a less clogged and longer life membrane
  • Leak-free Luer-Lok® and Luer-slip connections
  • Color-coded filters for easy organization
Prepare reliable aqueous, organic, and biomolecular samples with membrane filtration and a prefilter (depth filter) inside a polypropylene (PP) housing. The prefilter adds a filtration stage to remove larger particles which create unwanted buildup. Useful with solutions with a high viscosity like oils or high particulate samples.
Each housing comes with a color-coded retainer ring seal to prevent leaks and sample loss, and the color helps to visually show you the membrane type inside the housing. Syringe filters have Luer-Lok and Luer-slip connections for easy, quick and tight connections to tubing, syringes, or needles. Filter packs are available in a resealable storage container to prevent contamination during multiple opening and closings.
Call For Price
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Diameter (mm) Pore Size (µm) Description
Availability Pricing
COMPARE
0.22 Syringe Filter w/ Prefilter, Regenerated Cellulose, 25 mm dia., 0.22 µm; 100/pk
Call For Price
COMPARE
0.45 Syringe Filter w/ Prefilter, Regenerated Cellulose, 25 mm dia., 0.45 µm; 100/pk
Call For Price
COMPARE
0.45 Syringe Filter w/ Prefilter, Regenerated Cellulose, 30 mm dia., 0.45 µm; 100/pk