This product is now discontinued, View Related Products
Kinesis KX Regenerated Cellulose Filter Membranes
Remove particulate without adversely affecting your final product
  • Use membrane filtration for samples holding suspended solids
  • Highly efficient filtration of particles according to size
  • Complete sample filtration with the least sample loss and energy use
  • Filtration for HPLC sample preparation and mobile phase
  • Uniform manufacturing for repeatable results
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Diameter (mm) Pore Size (µm)
Availability Pricing
COMPARE
25 0.22
Call For Price
COMPARE
25 0.45