Kinesis® KX+ LCMS Certified High-Recovery Screw Top Glass Vials with Caps
Super clean for highly sensitive GCMS and LCMS applications
  • Vials and caps are lot tested by LCMS for cleanliness—certificate of analysis included, complete with MS scan
  • Minimal pH shift—vials and closures have a neutral pH factor
  • Compatible with all autosamplers
LCMS certified vials are manufactured using a process that reduces surface activity, improving analytical quantification and minimizing pH shifts in the samples. Surface metals such as sodium and boron that can contaminate samples and interfere with trace analysis are effectively removed. Screw vials feature 9-mm short thread with a unique thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
High-recovery vials deliver maximum sample capacity (1.5 mL) with minimal residual volume (<4 µL)—use with bottom draw-port needles.
16,800.00 - ₹20,200.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Septum Type Tube Color Volume (mL)
Availability Pricing
COMPARE
PTFE/silicone Clear 1.5
₹16,800.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Clear 1.5
₹17,700.00INR / Pkg of 100
COMPARE
PTFE/silicone Amber 1.5
₹18,600.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Amber 1.5
₹20,200.00INR / Pkg of 100