Cole-Parmer® KX+ LCGC Certified Short Thread Screw Top Fused-Insert and Total Recovery Vials with Caps
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Short thread vials have a wide opening (6.0 mm) with a unique 9-mm thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Glass vials are made of Type I, Class A 33-expansion borosilicate glass. Fused-insert vials are ideal for handling small-volume samples while delivering low residual volume (<8 µL). Total-recovery vials deliver maximum sample capacity (1.2 mL) with minimal residual volume (<1 µL)—use with bottom or side draw-port needles.
23,000.00 - ₹32,400.00INR / Pkg of 100
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Septum Type Special Features Volume (mL)
Availability Pricing
COMPARE
PTFE/silicone Fused Insert 0.3
₹23,000.00INR / Pkg of 100
COMPARE
PTFE/silicone/PTFE Fused Insert 0.3
₹27,700.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Fused Insert 0.3
₹25,400.00INR / Pkg of 100
COMPARE
PTFE/silicone Total-Recovery 1.2
₹26,100.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Total-Recovery 1.2
₹32,400.00INR / Pkg of 100