This product is now discontinued, View Related Products
Cole-Parmer® KX+ LCGC Certified Limited-Volume Short Thread Screw Top Polypropylene Vials with Caps
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Short thread vials have a wide opening (6.0 mm) with a unique 9-mm thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
Limited-volume vials are excellent for handling small-volume samples while delivering low residual volume (<8 µL for 300 µL vials; ,80 µL for 700 µL vials). Polypropylene (PP) is ideal for samples where glass is not appropriate; as with samples that adhere to glass.
5,100.00 - ₹6,800.00INR / Pkg of 100
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description Septum Type Volume (mL)
Availability Pricing
COMPARE
LCGC Limited-Volume Screw Top Vials, Polypropylene, 9 mm Short Thread, 0.3 mL, PTFE/Silicone Septum; 100/pk PTFE/silicone 0.3
₹5,100.00INR / Pkg of 100
COMPARE
LCGC Limited-Volume Screw Top Vials, Polypropylene, 9 mm Short Thread, 0.3 mL, Pre-Slit PTFE/Silicone Septum; 100/pk Pre-slit PTFE/silicone 0.3
₹6,800.00INR / Pkg of 100
COMPARE
LCGC Limited-Volume Screw Top Vials, Polypropylene, 9 mm Short Thread, 0.7 mL, PTFE/Silicone Septum; 100/pk PTFE/silicone 0.7
₹5,100.00INR / Pkg of 100
COMPARE
LCGC Limited-Volume Screw Top Vials, Polypropylene, 9 mm Short Thread, 0.7 mL, Pre-Slit PTFE/Silicone Septum; 100/pk Pre-slit PTFE/silicone 0.7
₹6,800.00INR / Pkg of 100