Cole-Parmer® KX+ LCGC Certified High-Recovery Short Thread Screw Top Glass Vials with Caps
Decrease your risk of failed analysis and sample reruns
  • Vials and caps are lot tested by HPLC for cleanliness—certificate of analysis included, complete with chromatogram
  • Compatible with all autosamplers
  • Minimal pH shift—vials and closures have a neutral pH factor
Certified vials are tested to confirm that the vials, cap, and septa have been properly manufactured, handled, and packaged for quality results. Short thread vials have a wide opening (6.0 mm) with a unique 9-mm thread design that allows you to screw on the caps and still use them in a robotic autosampler. The thread sits much higher on the vial allowing space between the bottom of the thread and the shoulder of the vial.
High-recovery vials deliver maximum sample capacity (1.5 mL) with minimal residual volume (<4 µL)—use with bottom draw-port needles. Clear glass vials are made of Type I, Class A 33-expansion borosilicate glass. Amber vials are made of Type I, Class B 51-expansion borosilicate glass and are ideal for light-sensitive samples.
23,800.00 - ₹32,800.00INR / Pkg of 100
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Septum Type Tube Color Volume (mL)
Availability Pricing
COMPARE
PTFE/Silicone/PTFE Clear 1.5
₹29,100.00INR / Pkg of 100
COMPARE
PTFE/silicone Amber 1.5
₹29,200.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Amber 1.5
₹32,800.00INR / Pkg of 100
COMPARE
PTFE/silicone Clear 1.5
₹23,800.00INR / Pkg of 100
COMPARE
Pre-slit PTFE/silicone Clear 1.5
₹26,600.00INR / Pkg of 100