Heraeus Noblelight Hollow Cathode Lamps, Indium
Achieve reliable atomic absorption results with high-performance lamps for your spectrometers
  • Carry out quicker start-up times
  • Reduce your operating costs with longer lifetime amp hours
  • Continuous light intensity for longer periods of time
  • Data-coded and uncoded versions are available
Heraeus Noblelight indium (In) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamp’s lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer®, Varian/Agilent, and Unicam systems.
70,400.00 - ₹81,200.00INR / Pkg of 1
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Material Compatible With Description
Availability Pricing
COMPARE
Indium Uncoded Hollow Cathode Lamp, indium, 37 mm, uncoded
₹74,500.00INR / Pkg of 1
COMPARE
Indium Unicam Hollow Cathode Lamp, indium, 37 mm, Unicam coded
₹81,200.00INR / Pkg of 1
COMPARE
Indium Varian Hollow Cathode Lamp, indium, 37 mm, Varian coded
₹70,400.00INR / Pkg of 1