Cole-Parmer® Headspace Glass Vials
Designed for use in headspace analyzers
  • Made from Type I Class 51 borosilicate glass
  • Select from 20 mm crimp neck or 18 mm screw neck vials
Type I Class 51 borosilicate glass provide excellent chemical resistance to various solutions. Vial design ensures full automation compatibility for headspace analyzers for both handling and needle positioning. Crimp and screw caps are available with various septum materials.
44,300.00 - ₹105,000.00INR / Pkg of 1000
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Cap Type Tube Color Volume (mL)
Availability Pricing
COMPARE
Crimp Neck Clear 5
₹44,300.00INR / Pkg of 1000
COMPARE
Crimp Neck Clear 5
₹44,300.00INR / Pkg of 1000
COMPARE
DIN Crimp Neck Clear 10
₹46,900.00INR / Pkg of 1000
COMPARE
DIN Crimp Neck Clear 10
₹46,900.00INR / Pkg of 1000
COMPARE
DIN Crimp Neck Amber 10
₹55,800.00INR / Pkg of 1000
COMPARE
Crimp Neck Clear 10
₹46,900.00INR / Pkg of 1000
COMPARE
Crimp Neck Clear 20
₹49,600.00INR / Pkg of 1000
COMPARE
Crimp Neck Amber 20
₹62,900.00INR / Pkg of 1000
COMPARE
DIN Crimp Neck Clear 20
₹51,000.00INR / Pkg of 1000
COMPARE
DIN Crimp Neck Amber 20
₹61,000.00INR / Pkg of 1000
COMPARE
DIN Crimp Neck Clear 20
₹49,600.00INR / Pkg of 1000
COMPARE
Crimp Neck Clear 20
₹58,400.00INR / Pkg of 1000
COMPARE
Crimp Neck Clear 50
₹105,000.00INR / Pkg of 1000
COMPARE
Screw Neck Clear 20
₹78,100.00INR / Pkg of 1000
COMPARE
Precision Thread Screw Neck Clear 20
₹62,500.00INR / Pkg of 1000
COMPARE
Precision Thread Screw Neck Amber 20
₹96,200.00INR / Pkg of 1000
COMPARE
Crimp Neck Clear 20
₹72,500.00INR / Pkg of 1000
COMPARE
Crimp Neck Clear 20
₹52,700.00INR / Pkg of 1000
COMPARE
Precision Thread Screw Neck Clear 10
₹55,400.00INR / Pkg of 1000
COMPARE
Precision Thread Screw Neck Amber 10
₹89,000.00INR / Pkg of 1000
Showing 1- 20 of 20