This product is now discontinued, View Related Products
Kinesis® Glass-Lined Polypropylene Collection Plates
Glass lining provides excellent chemical resistance
  • Constructed of glass-lined polypropylene
Collection plates are available with three well shapes: V-bottom wells let you completely retrieve well content; U-bottom wells optimize coating and washing; and flat wells maximize light transmission.
53,300.00 - ₹137,000.00INR / Pkg of 10
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Number of Wells Well Shape Well Volume (µL)
Availability Pricing
COMPARE
384 Square, U bottom 120
₹116,000.00INR / Pkg of 10
COMPARE
384 Square, U bottom 240
₹53,300.00INR / Pkg of 6
COMPARE
96 Round, flat bottom 370
₹100,000.00INR / Pkg of 10
COMPARE
96 Round, flat bottom 2400
₹97,900.00INR / Pkg of 10
COMPARE
96 Round, U bottom 300
₹82,400.00INR / Pkg of 10
COMPARE
96 Round, U bottom 1200
₹137,000.00INR / Pkg of 10
COMPARE
96 Round, V bottom 220
₹121,000.00INR / Pkg of 10
Showing 1- 7 of 7