KIMBLE® Scintillation Vials with Caps, Glass
Vial trays feature alphanumeric coordinates for fast, easy identification
  • Converted from Type I, 33 expansion borosilicate glass tubing
  • Five trays per case, shrink-wrapped to preserve cleanliness
These scintillation vials are made of made of low potassium borosilicate glass that conforms to ASTM guidelines to deliver optimal background counts and high UV transmission, making them ideal for solvent reduction. Multiple vial and cap configuration options are available for a wide range of applications. All vials include alphanumeric trays to track and organize samples.
44,700.00 - ₹89,500.00INR / Case of 1000
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Outside Diameter (mm) Septum Type Volume (mL)
Availability Pricing
DWK Life Sciences (Kimble) Scintillation Vials, Glass, Urea Cap with Cork-Backed Foil, 7 mL; 1000/Cs WW-08918-14 Mfr # 74502-7
COMPARE
17 Cork-backed Foil 7
₹89,500.00INR / Case of 1000
DWK Life Sciences (Kimble) Scintillation Vials, Glass, Polypropylene Cap with Pulp-Backed Foil, 7 mL; 1000/Cs WW-08918-16 Mfr # 74503-7
COMPARE
17 Pulp-backed Foil 7
₹86,600.00INR / Case of 1000
DWK Life Sciences (Kimble) Scintillation Vials, Glass, Polypropylene Cap with PE Foam Liner, 20 mL; 500/Cs WW-08918-46 Mfr # 74505-20
COMPARE
27 Polyethylene foam 20
₹49,600.00INR / Case of 500
DWK Life Sciences (Kimble) Scintillation Vials, Glass, Urea Cap with Cone PE Liner, 20 mL; 500/Cs WW-08918-48 Mfr # 74516-20
COMPARE
27 Cone-shaped Polyethylene 20
₹44,700.00INR / Case of 500