KIMAX® Wide-Mouth Class A Volumetric Flasks
Get easier sampling and aliquots
  • Choose from volumetric flask with PTFE or glass stopper
  • Autoclavable
Class A volumetric flasks are available with a PTFE or glass stopper. Constructed of glass with uniform wall construction provides superior strength and durability. These volumetric flasks have a wide mouth that allows easy access for pipettes. Easy identification is achieved with large, easy-to-read markings. Flat bottoms are ideal for precise stirring with standard stir bars. The 5- to 250-mL capacity flasks are calibrated to meet ASTM E288 standards.
31,100.00 - ₹99,500.00INR / Case of 6
20 variations of this product are available.
See All Product Options Below
20 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Cap Type Tolerance (mL)
Availability Pricing
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 5 mL;  6/CS WW-34558-30 Mfr # 92812F-5
COMPARE
5 PTFE stopper ±0.08
₹44,000.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 10 mL; 6/CS WW-34558-32 Mfr # 92812F-10
COMPARE
10 PTFE stopper ±0.08
₹47,500.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 20 mL; 6/CS WW-34558-34 Mfr # 92812F-20
COMPARE
20 PTFE stopper ±0.08
₹51,900.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 25 mL; 6/CS WW-34558-36 Mfr # 92812F-25
COMPARE
25 PTFE stopper ±0.08
₹51,000.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 50 mL; 6/CS WW-34558-38 Mfr # 92812F-50
COMPARE
50 PTFE stopper ±0.08
₹52,400.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 100 mL; 6/CS WW-34558-40 Mfr # 92812F-100
COMPARE
100 PTFE stopper ±0.10
₹63,800.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 200 mL; 6/CS WW-34558-42 Mfr # 92812F-200
COMPARE
200 PTFE stopper ±0.2
₹82,900.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 250 mL; 6/CS WW-34558-44 Mfr # 92812F-250
COMPARE
250 PTFE stopper ±0.2
₹82,900.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 1000 mL; 1/CS WW-34558-48 Mfr # 92812F-1000
COMPARE
1000 PTFE stopper ±0.3
₹31,100.00INR / Case of 1
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with PTFE Stopper, 2000 mL; 1/CS WW-34558-50 Mfr # 92812F-2000
COMPARE
2000 PTFE stopper ±0.5
₹46,400.00INR / Case of 1
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 5 mL; 6/CS WW-34558-60 Mfr # 92812G-5
COMPARE
5 Glass stopper ±0.08
₹44,000.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 10 mL; 6/CS WW-34558-62 Mfr # 92812G-10
COMPARE
10 Glass stopper ±0.08
₹47,500.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 20 mL; 6/CS WW-34558-64 Mfr # 92812G-20
COMPARE
20 Glass stopper ±0.08
₹51,900.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 25 mL; 6/CS WW-34558-66 Mfr # 92812G-25
COMPARE
25 Glass stopper ±0.08
₹51,000.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 50 mL; 6/CS WW-34558-68 Mfr # 92812G-50
COMPARE
50 Glass stopper ±0.08
₹52,400.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 100 mL; 6/CS WW-34558-70 Mfr # 92812G-100
COMPARE
100 Glass stopper ±0.10
₹63,800.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 200 mL; 6/CS WW-34558-72 Mfr # 92812G-200
COMPARE
200 Glass stopper ±0.2
₹82,900.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 500 mL; 6/CS WW-34558-76 Mfr # 92812G-500
COMPARE
500 Glass stopper ±0.2
₹99,500.00INR / Case of 6
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 1000 mL; 1/CS WW-34558-78 Mfr # 92812G-1000
COMPARE
1000 Glass stopper ±0.3
₹31,100.00INR / Case of 1
DWK Life Sciences (Kimble) Wide-Mouth Class A Volumetric Flask with Glass Stopper, 2000 mL; 1/CS WW-34558-80 Mfr # 92812G-2000
COMPARE
2000 Glass stopper ±0.5
₹45,500.00INR / Case of 1
Showing 1- 20 of 20