Kimax West condensers for distillations
Medium- length West condensers are ideal for refluxing and distillation assemblies. Design lessens condensate trapping.Full-length West condensers feature a stadard taper outer joint at he top and a standard taper inner drip joint at the bottom. High cooling efficiency is achieved with the increased flow rate of the cooling medium due to the design of the narrow annular space.
34,000.00 - ₹36,900.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Approx Condensing Area (cm²)
Availability Pricing
DWK Life Sciences (Kimble) Kimax West condenser, full length, 242 mm, 24/40 joint WW-34705-98 Mfr # 452000-2410
COMPARE
34
₹34,000.00INR / Each
DWK Life Sciences (Kimble) Kimax West condenser, full length, 342 mm, 24/40 joint WW-34706-00 Mfr # 452000-2420
COMPARE
68
₹35,100.00INR / Each
DWK Life Sciences (Kimble) Kimax West condenser, full length, 442 mm, 24/40 joint WW-34706-01 Mfr # 452000-2430
COMPARE
102
₹36,900.00INR / Each