KIMAX® Square Class A Volumetric Flasks
Save valuable space with square design
  • Constructed of low extractable borosilicate glass
  • All sizes cover 25-35% less area than conventional volumetric flasks
  • Designed with marking spots
These KIMAX® square volumetric flasks are designed to take up less area and pack more closely on shelves and in refrigerators or other storage cabinets. Due to the added glass weight and the square section, flasks also have greater stability. Calibrated “To Contain” and meet ASTM Spec E288 and Class A compliance requirements.
14,300.00 - ₹64,900.00INR / Pkg of 1
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL)
Availability Pricing
DWK Life Sciences (Kimble) Square Class A Volumetric Flask, 50 mL WW-34560-36 Mfr # 28040-50
COMPARE
50
₹14,500.00INR / Pkg of 1
DWK Life Sciences (Kimble) Square Class A Volumetric Flask, 100 mL WW-34560-38 Mfr # 28040-100
COMPARE
100
₹14,300.00INR / Pkg of 1
DWK Life Sciences (Kimble) Square Class A Volumetric Flask, 200 mL WW-34560-40 Mfr # 28040-200
COMPARE
200
₹34,100.00INR / Pkg of 1
DWK Life Sciences (Kimble) Square Class A Volumetric Flask, 250 mL WW-34560-42 Mfr # 28040-250
COMPARE
250
₹34,100.00INR / Pkg of 1
DWK Life Sciences (Kimble) Square Class A Volumetric Flask, 500 mL WW-34560-44 Mfr # 28040-500
COMPARE
500
₹42,400.00INR / Pkg of 1
DWK Life Sciences (Kimble) Square Class A Volumetric Flask, 2000 mL WW-34560-48 Mfr # 28040-2000
COMPARE
2000
₹64,900.00INR / Pkg of 1