KIMAX Soxhlet Extraction Condensers
Choose the condenser by type and sample volume
  • Allihn condenser (aka bulb or reflux condenser) better suited to basic laboratory applications
Freidrichs condenser (aka spiral cold finger condenser) better suited to heavy or difficult applications.
43,300.00 - ₹70,600.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Condenser Type Flask Joint Size
Availability Pricing
DWK Life Sciences (Kimble) Kimax Allihn Condenser; 125 mL; 1/Pk WW-99037-26 Mfr # 456000-0021
COMPARE
125 Allihn 34/45
₹43,300.00INR / Each
DWK Life Sciences (Kimble) Kimax Allihn Condenser; 250 mL; 1/Pk WW-99037-28 Mfr # 456000-0022
COMPARE
250 Allihn 45/50
₹52,600.00INR / Each
DWK Life Sciences (Kimble) Kimax Friedrich Condenser; 125 mL; 1/Pk WW-99037-32 Mfr # 456250-0021
COMPARE
125 Freidrich 34/45
₹66,300.00INR / Each
DWK Life Sciences (Kimble) Kimax Friedrich Condenser; 250 mL; 1/Pk WW-99037-34 Mfr # 456250-0022
COMPARE
250 Freidrich 45/50
₹70,600.00INR / Each