This product is now discontinued, View Related Products
Kimax Reflux Condensers
Dewar condensers are ideal for liquid nitrogen, dry ice/acetone or ice/alcohol mixtures. Provide rapid condensation of volatile components.
Reflux condensers are complete with an internal coil-type cold finger. outer joint at the top, inner drip joint at the bottom and two hose connectors at the top for water inlet and outlet.
Call For Price
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Approx Condensing Area (cm²)
Availability Pricing
DWK Life Sciences (Kimble) Kimax Dewar condenser, 24/40 joint, 45 mm H WW-34705-82 Mfr # 457500-2420
COMPARE
Call For Price
DWK Life Sciences (Kimble) Kimax Dewar condenser, 24/40 joint, 55 mm H WW-34705-84 Mfr # 457500-2425
COMPARE
Call For Price
DWK Life Sciences (Kimble) Kimax Reflux condenser, cold finger, 14/20 joint WW-34705-86 Mfr # 283000-0000
COMPARE
72
Call For Price
DWK Life Sciences (Kimble) Kimax Reflux condenser, cold finger, 19/22 joint WW-34705-88 Mfr # 283010-0000
COMPARE
Call For Price
DWK Life Sciences (Kimble) Kimax Reflux condenser, cold finger, 24/40 joint, 290 mm H WW-34705-90 Mfr # 457000-0125
COMPARE
115
Call For Price
DWK Life Sciences (Kimble) Kimax Reflux condenser, cold finger, 24/40 joint, 390 mm H WW-34705-92 Mfr # 457000-0225
COMPARE
229