KIMAX® RAY-SORB® Wide-Mouth Class A Volumetric Flasks with Glass Stoppers
Protect your light sensitive samples
  • Volumetric flask features Class A tolerances for enhanced accuracy
  • Autoclavable
These KIMAX® RAY-SORB® heavy-duty, wide-mouth volumetric flasks are amber to protect your light-sensitive solutions. The wide neck design allows up to 50mL pipette access. Flat bottom allows even stirring with standard stir bars. Each flask features large, permanent volume markings.
38,000.00 - ₹121,000.00INR / Case of 6
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (mL) Description
Availability Pricing
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 5 mL; 6/CS WW-34557-60 Mfr # 92822G-5
COMPARE
5 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 5 mL; 6/Cs
₹54,700.00INR / Case of 6
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 10 mL; 6/CS WW-34557-62 Mfr # 92822G-10
COMPARE
10 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 10 mL; 6/Cs
₹58,700.00INR / Case of 6
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 25 mL; 6/CS WW-34557-66 Mfr # 92822G-25
COMPARE
25 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 25 mL; 6/Cs
₹60,400.00INR / Case of 6
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 50 mL; 6/CS WW-34557-68 Mfr # 92822G-50
COMPARE
50 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 50 mL; 6/Cs
₹65,300.00INR / Case of 6
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 100 mL; 6/CS WW-34557-70 Mfr # 92822G-100
COMPARE
100 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 100 mL; 6/Cs
₹88,800.00INR / Case of 6
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 200 mL; 6/CS WW-34557-72 Mfr # 92822G-200
COMPARE
200 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 200 mL; 6/Cs
₹97,900.00INR / Case of 6
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 250 mL; 6/CS WW-34557-74 Mfr # 92822G-250
COMPARE
250 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 250 mL; 6/Cs
₹103,000.00INR / Case of 6
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 500 mL; 6/CS WW-34557-76 Mfr # 92822G-500
COMPARE
500 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 500 mL; 6/Cs
₹121,000.00INR / Case of 6
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 1000 mL; 1/CS WW-34557-78 Mfr # 92822G-1000
COMPARE
1000 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 1000 mL; 1/Cs
₹38,000.00INR / Case of 1
DWK Life Sciences (Kimble) Ray-Sorb Class A Volumetric Flask, Wide-Mouth, 2000 mL; 1/CS WW-34557-80 Mfr # 92822G-2000
COMPARE
2000 Ray-Sorb Glass Volumetric Flask, Wide-Mouth, 2000 mL; 1/Cs
₹55,000.00INR / Case of 1
Showing 1- 10 of 10